LC/MS Analysis of Cardiac Depressants on Ascentis® Express Biphenyl
LC/MS Analysis of Cardiac Depressants on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 3.0 mm, 2.7 µm particles (64066-U)
column temp. 35 °C
mobile phase 10mM ammonium formate (70:30 water:acetonitrile) SW pH 7.0 (not adjusted)
flow rate 0.6 mL/min
pressure 1363 psi (94 bar)
sample 300 ng/mL in 85:15,  water:methanol
injection 5 µL
detector MS ESI(+), TOF, XIC
Description
Featured Industry Clinical
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS