Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC-MS Analysis of Carotene Compounds on Ascentis® RP-Amide
LC-MS Analysis of Carotene Compounds on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 µm particles (565305-U)
column temp. 35 C
mobile phase acetonitrile
flow rate 0.2 ml/min
sample 10 mg/L in acetonitrile
injection 5 µl
detector MS, APCI(+), SIM
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products