LC/MS Analysis of Cocaine, Cocaethylene, Oxycodone, and Noroxycodone on Discovery® HS F5
LC/MS Analysis of Cocaine, Cocaethylene, Oxycodone, and Noroxycodone on Discovery® HS F5
Conditions
column Discovery HS F5, 5 cm x 2.1 mm I.D., 3 µm particles (567500-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate in water, pH 6.8; [B] 10mM ammonium formate in acetonitrile, pH 6.8; (10:90, A:B)
flow rate 0.3 mL/min
sample 200 ng/mL in 25:75, water:acetonitrile
injection 2 µL
detector ESI(+)
Description
Categories Analytical Chromatography, Forensics and Toxicology, Clinical, drugs of abuse, Drugs of Abuse
Featured Industry Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC