LC/MS Analysis of Conjugated Bile Acids on Ascentis® Express C18
LC/MS Analysis of Conjugated Bile Acids on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 4.6 mm I.D., 2.7 µm particles (53826-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate, pH 4 with acetic acid; [B] acetonitrile
gradient 25 to 40% B in 10 min
flow rate 1.0 mL/min (postcolumn split to MS)
sample 10 mg/L each in mobile phase A
injection 5 µL
detector MS, ESI(-), SIR
Description
Categories Analytical Chromatography, Clinical, organic acids, Organic Acids
Featured Industry Clinical