LC/MS Analysis of Cyanuric Acid on Ascentis® Express HILIC
LC/MS Analysis of Cyanuric Acid on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 µm particles (53934-U)
column temp. 35 °C
mobile phase 5 mM ammonium formate in 95:5 (v/v) acetonitrile:water
flow rate 0.2 mL/min
pressure 328 psi (22.6 bar)
sample 1 µg/mL in mobile phase
injection 2 µL
detector MS, ESI(-), SIR mode
Description
Categories Analytical Chromatography, Environmental Analysis, Pesticides & Herbicides, Organics, Chromatograms, HPLC Chromatograms
Featured Industry Chemicals and Industrial Polymers