Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of CYP2D6 Inhibitors on Ascentis® Express HILIC
LC/MS Analysis of CYP2D6 Inhibitors on Ascentis® Express HILIC
Conditions
column Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 50 C
mobile phase 5 mM ammonium formate in 2:98 (v/v) water:acetonitrile
flow rate 1.2 ml/min
injection 0.5 µl
detector ESI(+), m/z 100-400
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express HILIC for the analysis of CYP2D6 Inhibitors.
Categories Analytical Chromatography, Clinical, Clinical Enzymes, antidepressants, HPLC Applications Enzymes, Proteins, Antidepressants, proteins
Featured Industry Clinical
Life Science and Biopharma