LC/MS Analysis of Drugs of Abuse on Ascentis® Express RP-Amide
LC/MS Analysis of Drugs of Abuse on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate in water; [B] 10mM ammonium formate in acetonitrile; (75:25, A:B)
flow rate 0.2 mL/min
sample 500 ng/mL in water
injection 2 µL
detector ESI(+)
Description
Analysis Note This applic.ation demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in diet cola
Categories Analytical Chromatography, Forensics and Toxicology, Drugs of Abuse, drugs of abuse
Featured Industry Forensics and Toxicology