Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Equine Performance-Enhancing Dopants GABA and AICAR on Ascentis® Express HILIC
LC/MS Analysis of Equine Performance-Enhancing Dopants GABA and AICAR on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 C
mobile phase [A] 5 mM ammonium formate in acetonitrile:water (95:5); [B] 5 mM ammonium formate in acetonitrile:water (70:30)
gradient 0 to 70% B in 3 min, held for 1 min
flow rate 0.4 ml/min
sample 1 µg/mL in acetonitrile
injection 2 µl
detector MS, ESI+/ESI-
Description
Analysis Note This study focuses on the analysis of possible dopants used in equine racing, polar compounds requiring HILIC chromatographic separation. The rapid separation of four such compounds is shown here on an Ascentis Express HILIC column. The highest grade solvents provided clean, robust operation. Cerilliant ethyl-β-D-glucuronide CRM was used for reliable quantification.
Categories Analytical Chromatography, Neurotransmitters
Featured Industry Forensics and Toxicology
Veterinary
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS