LC-MS Analysis of Explosives on Ascentis® Express C18
LC-MS Analysis of Explosives on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 µm particles (53814-U)
column temp. 50 C
mobile phase [A] methanol; [B] water; (50:50, A:B)
flow rate 0.9 ml/min
sample 100 ng/mL in 50:50, methanol:water
injection 5 µl
detector MS: APCI(-)
Description
Categories Analytical Chromatography, Forensics and Toxicology, explosives, Explosives
Featured Industry Forensics and Toxicology