LC/MS Analysis of Fructose and Sucrose on SeQuant®ZIC-HILIC
LC/MS Analysis of Fructose and Sucrose on SeQuant®ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, 20 x 2.1 mm, 3.5 μm, 100 Å (1.50439.0001)
mobile phase  [A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
flow rate 0.5 mL/min
pressure 576 psi (4.0 MPa)
column temp.  ambient
detector Shimadzu LC-2010 Evolution, Detector
voltage: 1.6 kV (Heat block temp: 200 ºC; CDL
temp:200 ºC; Scan range: m/z 150-450;
SIM mode: m/z 240 (Fructose) and 403 (Sucrose))
injection: 1 μL
Description
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS