LC/MS Analysis of Illicit Bath Salts on Ascentis® Express HILIC, Set 3
LC/MS Analysis of Illicit Bath Salts on Ascentis® Express HILIC, Set 3
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium formate in acetonitrile; [B] water; (98:2, A:B)
flow rate 0.6 mL/min
pressure 267 psi (18.4 bar)
sample 500 ng/mL in acetonitrile
injection 2 µL
detector MS, ESI(+) TOF, Summed Ion Chromatogram
Description
Analysis Note HILIC mode operation on an Ascentis Express column provided baseline resolution of these bath salts, including isobaric compounds. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust; trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Forensics and Toxicology, drugs of abuse, Drugs of Abuse
Featured Industry Forensics and Toxicology