LC/MS Analysis of Isobaric Bath Salts Mephedrone, Buphedrone, N-Ethylcathinone and MDAI on Ascentis® Express HILIC
LC/MS Analysis of Isobaric Bath Salts Mephedrone, Buphedrone, N-Ethylcathinone and MDAI on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 µm particles (53934-U)
column temp. 35 °C
mobile phase 5 mM ammonium formate (98:2 acetonitrile:water)
flow rate 0.6 mL/min
pressure 1262 psi (87 bar)
sample 20 ng/mL
injection 1 µL
detector ESI(+), 178 m/z
Description
Categories Analytical Chromatography, Forensics and Toxicology, Drugs of Abuse, drugs of abuse
Featured Industry Forensics and Toxicology
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS