LC/MS Analysis of Melamine and Hydrolysis Products on Ascentis® Express HILIC
LC/MS Analysis of Melamine and Hydrolysis Products on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 µm particles (53934-U)
column temp. 35 °C
mobile phase 5 mM ammonium formate in 95:5 (v/v) acetonitrile:water
flow rate 0.2 mL/min
sample 1 mg/L in mobile phase
injection 2 µL
detector MS, ESI(+), full scan
Description
Categories Analytical Chromatography, Amines, Amides, Pesticides & Herbicides
Featured Industry Food and Beverages