Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2 in Urine following Liquid/Liquid Extraction
LC/MS Analysis of Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2 in Urine following Liquid/Liquid Extraction
Conditions
sample preparation 125 µl urine spiked at 125 ng/mL each enantiomer, add 1 mL diethyl ether, mix/vortex 30 min, centrifuge at 10,000 rpm for 10 min. remove 500 µl aliquot of organic (upper) layer, (evaporate to dryness under nitrogen at 55 C, reconstitute in 500 µl mobile phase, filter through 0.22 µm PVDF membrane (Z227498))
column Astec CHIROBIOTIC V2, 15 cm x 4.6 mm I.D., 5 µm particles (15023AST)
column temp. 20 C
mobile phase [A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate 1.0 ml/min
pressure 1220 psi (84 bar)
injection 2.0 µl
detector MS, ESI(+),SIR m/z 150.1
Description
Analysis Note 125 μL urine spiked at 125 ng/mL each enantiomer, add 1 mL diethyl ether, mix/vortex 30 min, centrifuge at 10,000 rpm for 10 min. remove 500 μL aliquot of organic (upper) layer, evaporate to dryness under nitrogen at 55 °C, reconstitute in 500 μL mobile phase, filter through 0.22 μm PVDF membrane (Z227498)
Categories Analytical Chromatography, Forensics and Toxicology, enantiomers, drugs of abuse, Enantiomers, Drugs of Abuse
Featured Industry Forensics and Toxicology
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS