LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express HILIC
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express HILIC
Conditions
column Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 C
mobile phase [A] 5 mM ammonium formate in water, pH 6.5; [B] 5 mM ammonium formate in acetonitrile, pH 6.5; (2:98, A:B)
flow rate 0.3 ml/min
sample 1 mg/L each in mobile phase
injection 0.1 µl
detector ESI(+), m/z 50-400
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express HILIC for the analysis of omeprazole metabolites.
Categories Analytical Chromatography, Clinical, antiulcer compounds, Antiulcer Compounds
Featured Industry Clinical