Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express RP Amide (pH 3.4)
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express RP Amide (pH 3.4)
Conditions
column Ascentis Express RP-Amide, 5 cm x 2.1 mm I.D., 2.7 µm particles (53911-U)
column temp. 35 C
mobile phase [A] 10 mM ammonium formate in water, pH 3.4; [B] 10 mM ammonium formate in methanol, pH 3.4; (65:35, A:B)
flow rate 0.3 ml/min
sample 1 mg/L in mobile phase
injection 1 µl
detector ESI(+), m/z 50-400
Description
Analysis Note This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of omeprazole metabolites.
Categories Analytical Chromatography, Clinical, antiulcer compounds, Antiulcer Compounds
Featured Industry Clinical