LC/MS Analysis of Peptides, Basic, on Ascentis® C18
LC/MS Analysis of Peptides, Basic, on Ascentis® C18
Conditions
column Ascentis C18, 10 cm x 2.1 mm I.D., 5 µm particles (581326-U)
column temp. 22 °C
mobile phase [A] 20 mM formic acid in water; [B]: 50:50 (v/v) 20 mM formic acid in water:20 mM formic acid in acetonitrile
gradient 20 to 60% B in 20 min
flow rate 0.2 mL/min
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Peptides