Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Pesticides on Ascentis® C18 (5 cm x 2.1 mm x 3 μm)
LC/MS Analysis of Pesticides on Ascentis® C18 (5 cm x 2.1 mm x 3 μm)
Conditions
column Ascentis C18, 5 cm x 2.1 mm I.D., 3 µm particles (581300-U)
column temp. 35 C
mobile phase [A] 0.1% ammonium acetate, pH unadjusted; [B] acetonitrile
gradient 10 to 80% B in 10 min
flow rate 0.2 ml/min
sample 1 µg/mL each in 90:10 water:acetonitrile
injection 5 µl
detector MS, (+) ESI, Selected Ion Recording Mode
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Environmental Analysis, Pesticide and Metabolite Residues, Organics, Pesticides & Herbicides, Chromatograms, Applications (Chromatograms), HPLC Chromatograms
Featured Industry Environmental