LC/MS Analysis of Phosphatidylethanol Metabolites on Ascentis® Express C18
LC/MS Analysis of Phosphatidylethanol Metabolites on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.0 µm particles (50811-U)
column temp. 50 °C
mobile phase 5 mM ammonium formate in [A] water; [B] methanol; [C] acetonitrile; (5:47.5:47.5, A:B:C)
flow rate 0.5 mL/min
pressure 2760 psi (190 bar)
sample 500 ng/mL in 50:50, water:acetonitrile
injection 2 µL
detector MS, ESI-, combined SIR: 699.7, 701.7 m/z
Description
Analysis Note Phosphatidylethanol and its metabolites in blood are markers of ethanol consumption. Shown here is the rapid resolution of the metabolites on an Ascentis® Express C18 column.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Forensics and Toxicology
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS