LC/MS Analysis of Pilocarpine and Betaxolol in the Presence of Benzalkonium Chlorides on Ascentis® Express HILIC
LC/MS Analysis of Pilocarpine and Betaxolol in the Presence of Benzalkonium Chlorides on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 °C
mobile phase [A] 2 mM arrmonium acetate, pH 6.7 in [B] 10:90 (v/v) water:acetonitrile
flow rate 1.0 mL/min
pressure 1160 psi (80 bar)
sample 10 µg/mL each in acetonitrile
injection 2 µL
detector ESI(+), XlC, m/z 304, 332 (BACs), m/z 209 (pilocarpine), m/z 308 (betaxolol)
Description
Categories Analytical Chromatography, Beta-blockers, Cholinergic agonist