Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Polar Amino Acids on Ascentis® Si (HILIC Mode)
LC/MS Analysis of Polar Amino Acids on Ascentis® Si (HILIC Mode)
Conditions
column Ascentis Si, 10 cm x 2.1 mm I.D., 5 µm particles (56403AST)
column temp. 35 C
mobile phase [A] 100 mM ammonium formate, pH 3.0 with formic acid; [B] water; [C] acetonitrile
gradient 5% A constant; 10 to 16% B, 85 to 79% C in 20 min
flow rate 0.3 ml/min
sample 0.1 g/L each in mobile phase
injection 2 µl
detector ESI(+)
Description
Categories Analytical Chromatography, Amino Acids