LC/MS Analysis of Sudan Red Dyes on Ascentis® RP-Amide
LC/MS Analysis of Sudan Red Dyes on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in water (pH unadjusted): [B] acetonitrile; (15:85, A:B)
flow rate 1 mL/min, split to the MS
sample 1 µg/mL in 30:70, 0.1% formic acid in water (pH unadjusted):acetonitrile
injection 15 µL
detector MS, APCI (+) in selected ion recording (SIR) mode
Description
Categories Analytical Chromatography, Flavors, Fragrances
Featured Industry Chemicals and Industrial Polymers
Food and Beverages