LC/MS Analysis of the Plant Growth Regulators Chlormequat and Mepiquat on SeQuant® ZIC-HILIC
LC/MS Analysis of the Plant Growth Regulators Chlormequat and Mepiquat on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 2.1 mm, 3.5 μm, 200 Å (1.50447.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate 0.2 mL/min
column temp. ambient
detector electrospray-MS in postive mode (ESI+) (single ion monitoring (SIM) at m/z 114 and 122)
injection: 20 μL
Description
Featured Industry Agriculture
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS