LC/MS Analysis of thyroid hormones on Titan™ PFP
LC/MS Analysis of thyroid hormones on Titan™ PFP
Conditions
column Titan PFP, 5 cm X 3.0 mm I.D., 120Ã…, 1.9 µm particles (569682-U)
column temp. 45 °C
mobile phase [A] 100 mM ammonium acetate, pH 4.0 with acidic acid: water (10:90); [B] 100 mM ammonium acetate, pH with acidic acid: acetonitrile (10:90)
gradient 15 to 55% B in 3 min; to 100% B in 0.1 min; held at 100% B for 1 min; to 15% B in 0.2 min; held at 15% B for 1.7 min
flow rate 0.5 mL/min
pressure 1462 psi (101 bar)
sample 100 ng/mL in 99:1 water:acetonitrile
injection 4 µL
detector MS, ESI (+) TOF, SIM, (T3 and rT3 m/z 651.8000; T4 m/z 777.7000
Description
Featured Industry Life Science and Biopharma
General description The thyroid hormones, triiodothyronine (T3) and its prohormone, thyroxine (T4), are tyrosine-based hormones produced by the thyroid gland that are primarily responsible for regulation of metabolism. Thyroxine, or T4, is the thyroid "storage hormone."Triodothyronine, or T3, is the "energy hormone." For the body to use T4, it must first convert it to the active hormone T3, giving energy to every cell in the body. Another critical thyroid hormone is reverse T3 (rT3). Reverse T3 is the body’s “emergency brake.” rT3 is a metabolite of T4 (thyroxine). Typically, when T4 loses an atom of iodine -- a process known as monodeiodination, or T4 to T3 conversion -- it becomes triiodothyronine (T3), the active thyroid hormone. But in some cases, the body conserves energy by converting the T4 instead into rT3, an inactive form of T3 that is incapable of delivering oxygen and energy to the cells, as T3 does. The Titan PFP shows baseline resolution of these three common thyroid hormones in a 6 minute gradient.
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS