LC/MS Analysis of Vaccinium Antioxidants on Ascentis® Express C18
LC/MS Analysis of Vaccinium Antioxidants on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm X 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in 75:25 (v/v) acetonitrile:water; (78:22, A:B)
flow rate 0.3 mL/min
pressure 3260 psi (225 bar)
sample 20 µg/mL in 0.1% formic acid
injection 5 µL
detector ESI(+), TOF
Description
Analysis Note Fruits of the genus Vaccinium are popular sources for anthocyanidin antioxidants. This genus includes the commonly known blueberry, huckleberry, and bilberry.
Categories Analytical Chromatography, Antioxidants
Featured Industry Vitamins, Nutraceuticals, and Natural Products