Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Extracted Ion Chromatogram of MSX, Sulfone, and Sulfoxide on Ascentis® Express OH5 (HILIC Mode)
LC/MS Extracted Ion Chromatogram of MSX, Sulfone, and Sulfoxide on Ascentis® Express OH5 (HILIC Mode)
Conditions
column Ascentis Express OH5, 10 cm x 3 mm I.D., 2.7 µm particles (53769-U)
column temp. ambient
mobile phase [A] 0.1% formic acid, pH 3.5 with ammonium formate; [B] acetonitrile; (25:75, A:B)
flow rate 0.4 ml/min
pressure 1350 psi (93 bar)
sample 10 µg/mL in 90% methanol
injection 5 µl
detector ESI (+), scan m/z 150-300
Description
Analysis Note This figure shows the extracted ion currents for methionine sulfoximine-related compounds on Ascentis Express OH5. The OH5 shows excellent peak shape compared to traditional HILIC on bare silica.
Categories Analytical Chromatography, Sulfur compounds, Amino Acids
Featured Industry Chemicals and Industrial Polymers