Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS/MS Analysis of Citrinin on Ascentis® Express Phenyl-Hexyl
LC/MS/MS Analysis of Citrinin on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 5 cm x 2.1 mm I.D., 2.7 µm particles (53334-U)
column temp. 40 C
mobile phase [A] 5mM ammonium acetate with 0.1% formic acid in water, [B] 5 mM ammonium acetate with 0.1% formic acid in methanol
gradient 50% to 95% B in 2 min, held at 95% B for 1 min, to 50% B in 0.1 min, held at 50% B for 0.9 min
flow rate 0.4 ml/min
pressure 3263 psi (225 bar)
sample acetonitirle:water (50:50)
injection 2 µl
detector MS/MS ESI(+), MRM, 251.1 > 233.1 (Citrinin), 264.1 > 246.1 (13C-Citrinin)
Description
Analysis Note Using Ascentis Express Phenyl-Hexyl, a fast rugged analytical method was established for citrinin.
Categories Analytical Chromatography, Mycotoxins
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS