LC/MS/MS Analysis of Marine Toxins in Shellfish According to EU Regulations on Ascentis® Express C18
LC/MS/MS Analysis of Marine Toxins in Shellfish According to EU Regulations on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2 µm particles (50813-U)
column temp. 40 °C
mobile phase [A] 2mM ammonium acetate with 0.1% formic acid in acetonitrile:water (5:95), [B] 2 mM ammonium acetate with 0.1% formic acid in acetonitrile:water (95:5)
gradient 20% to 80% B in 4 min, to 95% B in 1 min, held at 95% B for 1 min, to 20% B in 0.5 min, held at 20% B for 1.5 min
flow rate 0.6 mL/min
pressure 5801 psi (400 bar)
injection 10 µL
detector MS/MS ESI(+) and ESI(-), MRM (see Figure for transitions)
Description
Analysis Note Biotoxins in shellfish have recently come under increased scrutiny. The European Union (EU) has issued regulation changes requiring all shellfish commodities to be tested for levels of these toxins. Shown here is a rapid and reproducible method for the extraction and analysis of lipophilic marine toxins from shellfish. Separation was achieved in less than seven minutes on an Ascentis® Express C18 column. Recoveries ranged from 80-120% using matrix-matched calibration standards. Certified reference materials provided reliable quantification.
Categories Analytical Chromatography, Natural products
Featured Industry Forensics and Toxicology
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes mussel tissue homogenate matrix CRM (National Research Council Canada, part no. CRM-ZERO-Mus) extracted with methanol
suitability application for LC-MS