LC/MS/MS Analysis of Testosterone and Metabolites on Ascentis® Express C18
LC/MS/MS Analysis of Testosterone and Metabolites on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 µm particles (53822-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium formate water pH 4.0 formic acid; [B] 0.1% formic acid acetonitrile
gradient 10 to 70% B in 5 min; held at 70% B for 3 min
flow rate 0.6 mL/min
pressure 3698 psi (255 bar)
sample 1000 ng/mL in 40:60 (water:methanol)
injection 5 µL
detector MS, ESI(+), TIC, m/z 100-1000
Description
Analysis Note An Ascentis Expres F5 Column provided rapid, baseline resolution of testosterone and metabolites. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Clinical, Steroids, steroids
Featured Industry Clinical