LC/MS (TOF) Analysis of Antiarrhythmic Drugs and Metabolites in Plasma on Ascentis® Express HILIC after Sample Prep using HybridSPE®-Phospholipid
LC/MS (TOF) Analysis of Antiarrhythmic Drugs and Metabolites in Plasma on Ascentis® Express HILIC after Sample Prep using HybridSPE®-Phospholipid
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix rat plasma stabilized with K2EDTA was spiked directly from stock standard to a level of 400 ng/mL
SPE tube/cartridge HybridSPE-Phospholipid 96-Well Plate, 50 mg/2 mL (575656-U)
sample addition apply 100 µL of plasma to plate, followed by 300 µL of 1% formic acid acetonitrile. Agitate via vortex for 4 min
elution place on vacuum manifold and apply 10" Hg vacuum for 4 minutes. Collect filtrate and analyze directly.
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 µm particles (53939-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in acetonitrile; (5:95, A:B, pH 7.0 with formic acid)
flow rate 0.4 mL/min
pressure 1305 psi (90 bar)
sample plasma extract, analyte concentration of final sample work up is equivalent to 100 ng/mL
injection 0.5 µL
detector ESI(+), full scan, m/z 100-1000
Description
Analysis Note The basic characteristics of these compounds make them targets for HILIC chromatographic separation. HILIC mobile phases consist of a high composition of acetonitrile, which facilitates the direct analysis of precipitated plasma samples without the need for additional sample solvent exchange. In most cases, the high organic mobile phase also facilitates increased analyte response in ESI(+) MS detections. This application used HybridSPE-Phospholipid sample prep to remove phospholipids and precipitated proteins prior to LC/MS analysis on an Ascentis Express HILIC Fused-Core HPLC column. The highest grade solvents provided clean, robust operation. Cerilliant CRMs provided reliable quantification. The resulting method was robust and sensitive.
Categories Analytical Chromatography, SPE Applications, Clinical, Pharmaceutical Analysis, antiarrhythmic drugs, cardiac drugs, Antiarrhythmic drugs, Cardiac Drugs, Cardiac drugs, Antiarrythmic
Featured Industry Clinical
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, HybridSPE is a registered trademark of Sigma-Aldrich Co. LLC