LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC
LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 µm particles (53934-U)
column temp. 55 °C
mobile phase 2 mM ammonium formate in 90:10 (v:v) acetonitrile:water
flow rate 0.3 mL/min
sample 400 ng/mL in acetonitrile
injection 2 µL
detector TOF/MS
Description
Analysis Note The rapid analysis by HPLC with TOF-MS detection of seven opioids is shown here using an Ascentis Express HILIC column. Cerilliant CRMs were used to ensure reliable MS identification and quantification.
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, drugs of abuse, opiate, Drugs of Abuse
Featured Industry Clinical Testing
Forensics and Toxicology