Mobile Phase Optimization in the Analysis of Lysozyme on Zenix® SEC-300
Mobile Phase Optimization in the Analysis of Lysozyme on Zenix® SEC-300
Conditions
column Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 µm particles (Z777028)
mobile phase as indicated on figure
flow rate 0.35 ml/min
temp. 25 C
sample lysozyme, 1 mg/mL
injection 5 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, HPLC Applications Enzymes
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies