Monitoring the Formation of PEGylated Protein (ß-Lactoglobulin) Reaction Products by HPLC using TSKgel® SP-STAT
Monitoring the Formation of PEGylated Protein (ß-Lactoglobulin) Reaction Products by HPLC using TSKgel® SP-STAT
Conditions
column TSKgel SP-STAT, 3.5 cm x 3.0 mm I.D., 10 µm particles (821963)
mobile phase [A] 20 mM sodium acetate buffer, pH 5.0; [B] 20 mM sodium acetate buffer, 1 M NaCl pH 5.0
gradient 0 to 100% B in 2 min
flow rate 2 mL/min
sample PEGylated ß-lactoglobulin
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.