Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
NP-HPLC with Fluorimetric Detector of Tocopherols in Extra Virgin Olive Oil Samples on Ascentis® Si
NP-HPLC with Fluorimetric Detector of Tocopherols in Extra Virgin Olive Oil Samples on Ascentis® Si
Conditions
column Ascentis Si, 10 cm x 1.0 mm I.D., 3 µm particles (581520-U)
mobile phase [A] hexane; [B] isopropanol; (99:1; A:B)
flow rate 50 µL/min
detector fluorescence, ex: 290 nm, em: 330 nm
Description
Analysis Note Sample preparation:
Extra virgin olive oil
10 mg diluted in 1 mL of n-hexane
Extra virgin olive oil by-products
100 g of each sample was extracted with 100 mL of n-hexane 3 times, n-hexane fractions were collected and brought to dryness in a rotary evaporator, 10 mg of the residue was diluted in 1 mL of n-hexane prior to analysis
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability suitable for HPLC