Protein Molecular Weight Calibration Curve on Zenix® Size Exclusion Columns, Effect of Pore Size
Protein Molecular Weight Calibration Curve on Zenix® Size Exclusion Columns, Effect of Pore Size
Conditions
column Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 µm, 150 Angstrom particles (Z777018)
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm, 300 Angstrom particles (Z777033)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1 ml/min
sample each 1 mg/mL except uracil, 2.5 mg/mL
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies