Protein MW Calibration Curves on SRT® SEC-100, -150 and -300, 30 cm x 7.8 mm I.D., 5 μm particles
Protein MW Calibration Curves on SRT® SEC-100, -150 and -300, 30 cm x 7.8 mm I.D., 5 μm particles
Conditions
column SRT(R) SEC-100, -150 and -300, 30 cm x 7.8 mm I.D., 5 µm particles (Z777039)
mobile phase 150 mM potassium phosphate, monobasic, pH 7 (adjusted with potassium phosphate)
flow rate 1 ml/min
injection 10 µl
detector UV, 214 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies