Reproducibility of Semi-Preparative Separations on SeQuant® ZIC-HILIC
Reproducibility of Semi-Preparative Separations on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 10 mm, 5 μm, 200 Å (1.50493.0001)
mobile phase [A] acetonitrile; [B] 25 mM ammonium acetate, pH 6.8; (80:20, A:B)
flow rate 2.2 mL/min
pressure 432 psi (3.0 MPa)
detector 254 nm
injection: 100 μL
Description
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany