Retention vs Particle Porosity (100 and 200 Å) on SeQuant ZIC-HILIC
Retention vs Particle Porosity (100 and 200 Å) on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 2.1 mm, 3.5 µm, 200 Ã… (1.50448.0001)
column temp. 30 C
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
flow rate 0.1 ml/min
injection 5 µl
detector 254 nm
Description
Featured Industry General Analytical