RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl
RP-HPLC Analysis of Benzodiazepines on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10 cm x 3.0 mm I.D., 2.7 µm particles (64066-U)
column temp. 35 °C
mobile phase [A] Water; [B] acetonitrile; (70:30, A:B)
flow rate 1.0 mL/min
pressure 5294 psi (365 bar)
sample Benzodiazepine multicomponent Mixture-8 diluted to 50ug/mL in 50:50 Water:Methanol
injection 1.0 µL
detector UV, 254 nm
Description
Analysis Note This analysis demonstrates the applicability of the Ascentis Express Biphenyl stationary phase for the analysis of benzodiazepines, a group of psychoactive drugs that act as GABA agonists. The stationary phase provides alternative selectivity and improved resolution to existing phenyl chemistires.
Categories Analytical Chromatography, Benzodiazepines