RP-HPLC Analysis of Substituted Phenethylamine Psychedelics on Ascentis® Express 2.7 μm RP-Amide
RP-HPLC Analysis of Substituted Phenethylamine Psychedelics on Ascentis® Express 2.7 μm RP-Amide
Conditions
column Ascentis Express RP-Amide 10cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 20 mM Ammonium acetate pH 4.0 with Acetic  Acid; [B] ACN; (85:15, A:B)
gradient 15% B for 2 min; 15 to 30% B in 6 min, held at 30% B for 4 min
flow rate 0.6 mL/min
pressure 4757 psi (328 bar)
sample 25C-NBOMe HCl, 25B-NBOMe HCl, 25I-NBF HCl, 25INBMD HCl, 25I-NBOME HCl, 20 µg/mL in 90:10 mobile phase A:mobile phase B
injection 5 µL
detector UV, 280 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the HPLC separation of substituted phenethylamine psychedelics (Bath Salts). This application shows the use of UV detection for this set of compounds.
Categories Analytical Chromatography, Drugs of Abuse