Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
SFC Analysis of Metalaxyl Enantiomers on Astec® Cellulose DMP
SFC Analysis of Metalaxyl Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 35 C
mobile phase 20% methanol in CO2
flow rate 3 ml/min
pressure 1450 psi (100 bar)
sample 1 mg/mL in methanol
injection 5 µl
detector UV, 220 nm
Description
Analysis Note Metalaxyl is a 1:1 mixture of (R)- and (S)-2-[(2,6-dimethylphenyl)methoxyacetylamino]propionic acid methyl ester. Both enantiomers are fungicides used in agriculture, horticulture, and forestry. They act by inhibiting mycelial growth and spore formation. Metalaxyl-M is the biologically active (R)-enantiomer. Technical-grade metalaxyl-M consists of a miniµm of 97% of the (R)-enantiomer and 3% of the (S)-enantiomer. Chiral LC on Astec Cellulose DMP was used to separate the two enantiomers.
Categories Analytical Chromatography, SFC Applications
Featured Industry Agriculture
Environmental
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SFC, application for HPLC