UHPLC Analysis of a Rabbit Secretory IgA Monoclonal Antibody on BIOshell™ A400 Protein C4
UHPLC Analysis of a Rabbit Secretory IgA Monoclonal Antibody on BIOshell™ A400 Protein C4
Conditions
column BIOshell A400 Protein C4; 10 cm x 2.1 mm I.D., 3.4 µm particles (66825-U)
column temp. 75 °C
mobile phase [A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile
gradient [A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile
flow rate 0.2 mL/min
pressure 550 psi (38 bar)
sample Rabbit secretory IgA, 100 µg/mL, 0.05% TFA in water
injection 1 µL
detector UV, 215 nm
Description
Analysis Note Secretory immunoglobulin A (IgA) exists as a complex of 2-4 IgA monomers linked through a 15 kDa polypeptide J chain. Rabbit secretory IgA is a 385 kDa antibody composed of two IgA monomers and the J chain. Secretory IgA is resistant to degradative enzymes; therefore, it plays a key role in providing protection against microbes that µLtiply in bodily secretions. This antibody was chromatographically analysed on a BIOshell A400 Protein C4 column. A prominent shoulder peak was partially resolved from the main analyte peak, indicating the presence of impurities and/or other, post-translationally modified variants.
Categories Analytical Chromatography, Antibodies, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC