Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Antihistamines on Ascentis® Express C18
UHPLC Analysis of Antihistamines on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 3.0 mm I.D., 5 µm particles (50526-U)
column temp. 40 C
mobile phase [A] 0.02M sodium phosphate, pH 2.6; [B] methanol
gradient 50% B for 0.5 min; to 75% B in 2 min; held for 2 minutes
flow rate 1.0 ml/min
pressure 2770 psi (191 bar)
sample Dissolved in 20:80 (v:V0 water:methanol
injection 1.0 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Antihistamines
Featured Industry Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC