UHPLC Analysis of Boswellic Acids on Titan™ C18
UHPLC Analysis of Boswellic Acids on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 35 °C
mobile phase [A] 0.1% phosphoric acid; [B] acetonitrile; (10:90, A:B)
flow rate 0.4 mL/min
pressure 2450 psi (169 bar)
sample 6 Boswellic Acids 50µg/mL in 70:30, water:methanol
injection 2 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC