UHPLC Analysis of Deuterated Isomers on Titan™ C18
UHPLC Analysis of Deuterated Isomers on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (50:50, A:B)
flow rate 0.4 mL/min
pressure 5650 psi (390 bar)
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Anticonvulsant/antiepiletic/antiseizure, Aromatics
Featured Industry Chemicals and Industrial Polymers
Petroleum
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC