Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Diazepam and Metabolites on Titan™ C18, Competitive Comparison
UHPLC Analysis of Diazepam and Metabolites on Titan™ C18, Competitive Comparison
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 35 C
mobile phase [A] 0.1% formic acid; [B] acetonitrile; (60:40, A:B)
pressure 6450 psi (445 bar)
detector UV, 254 nm
Description
Analysis Note This application shows a comparison between Titan C18 and a competitive column for diazepam and its metabolites using a MS compatible mobile phase.
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Drugs of Abuse, antiepileptic/antiseizure, antidepressants, Antidepressants, drugs of abuse
Featured Industry Clinical
Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
Other Notes Used 75 μm I.D. x 250 mm L tubing in system.
suitability application for UHPLC