UHPLC Analysis of Fat Soluble Vitamins on Titan™ C18, Acetonitrile Mobile Phase
UHPLC Analysis of Fat Soluble Vitamins on Titan™ C18, Acetonitrile Mobile Phase
Conditions
column TITAN C18, 50 mm x 2.1mm I.D. x 1.9 µm particles (577122-U)
column temp. 30 C
mobile phase 100% acetonitrile
flow rate 0.4 ml/min
pressure 1755 psi (121 bar)
sample All compounds at 0.5 mg/mL except retinyl acetate at 0.05 mg/mL in methanol
injection 0.5 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC