UHPLC Analysis of Flakka Compounds on Ascentis® Express HILIC
UHPLC Analysis of Flakka Compounds on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2 µm (51409-U)
column temp. 35 °C
mobile phase 10 mM ammonium formate in acetonitrile:water (95:5)
flow rate 0.4 mL/min
sample 50 µg/mL in methanol:acetonitrile (10:90)
injection 1 µL
detector UV, 265 nm
Description
Analysis Note The rapid separation of the street drug Flakka (α-PVP) and related compounds is shown here on an Ascentis Express HILIC column. Highest grade UHPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Forensics and Toxicology