Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Haloperidol and Metabolite on Titan™ C18
UHPLC Analysis of Haloperidol and Metabolite on Titan™ C18
Conditions
column Titan C18 5 cm x 2.1 mm, 1.9 µm particles (577122-U)
column temp. 35 C
mobile phase [A] 10 mM potassium phosphate, monobasic pH 3.0; [B] acetonitrile; (70:30, A:B)
flow rate 0.6 ml/min
pressure 4310 psi (297 bar)
sample 1-10 µg/mL in 70:30, water:methanol
injection 1 µl
detector UV, 200 nm
Description
Categories Analytical Chromatography, Antipsychotics
Featured Industry Life Science and Biopharma
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC
Materials
Related Links
Categories