Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Nevirapine and Imiquimod on Titan™ C18
UHPLC Analysis of Nevirapine and Imiquimod on Titan™ C18
Conditions
column TITAN C18, 10 cm x 2. 1 mm I.D., 1.9 um particles (577124-U)
column temp. 35 C
mobile phase [A] 10 mM ammonium acetate, pH 7.0; [B] acetonitrile (70:30, A;B)
flow rate 0.3 ml/min
sample 300 ng/mL each in acetonitrile:water (3:1)
injection 2 µl
detector MS, XIC m/z 267.12 and 241.15
Description
Categories Analytical Chromatography, Antiviral
Featured Industry Pharmaceutical (small molecule)
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC